Assay Details
Gene Symbol: LOC101928591 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135065.1 | 6 - 7 | 866 | 93 | ||
GenBank mRNA | AY726565.1 | - | 6 - 7 | 829 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1499 | LINC01499 | -202.6kb | Anti-sense | Chr. 11 - 41714568 - 41836442 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC100507205 | LOC100507205 | 126.3kb | Sense | Chr. 11 - 42187743 - 42253690 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
