Assay Details
| Gene Symbol: RPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB209803.1 | - | 1 | 152 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4754 | MIR4754 | mir-4754 | -6.2kb | Anti-sense | Chr. 19 - 58386770 - 58386858 on NCBI Build GRCh38 |
| Downstream | ring finger protein 225 | RNF225 | 855.0 | Sense | Chr. 19 - 58395081 - 58398954 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000598807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000599909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000196551.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000596314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000598807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000599909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | ENST00000601521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | NM_001009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009045.hg.1 | NM_001009.3 | Refseq |

RPS5
Genomic Map