Assay Details
| Gene Symbol: PTCSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK023948.1 | - | 1 | 2652 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 7848 | MIR7848 | hsa-mir-7848 | -8.4kb | Anti-sense | Chr. 8 - 133046481 - 133046581 on NCBI Build GRCh38 |
| Downstream | WNT1 inducible signaling pathway protein 1 | WISP1 |
CCN4, WISP1c, WISP1i, WISP1tc |
133.3kb | Sense | Chr. 8 - 133191039 - 133231690 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

PTCSC1
Genomic Map