Assay Details
| Gene Symbol: NRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017005186.1 | XP_016860675.1 | 17 | 6200 | 99 | |
| GenBank mRNA | AK024680.1 | - | 1 | 1550 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | par-3 family cell polarity regulator beta | PARD3B |
ALS2CR19, PAR3B, PAR3L, PAR3beta |
-157.7kb | Sense | Chr. 2 - 204545793 - 205620162 on NCBI Build GRCh38 |
| Downstream | INO80 complex subunit D | INO80D | 214.0kb | Anti-sense | Chr. 2 - 205993721 - 206086182 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NRP2
Genomic Map