Assay Details
Gene Symbol: C17orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113434.4 | NP_001106905.1 | 2 | 7741 | 95 | |
XM_006721514.2 | XP_006721577.1 | 3 | 1417 | 95 | ||
GenBank mRNA | AK096256.1 | - | 1 | 1653 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | potassium voltage-gated channel subfamily J member 12 | KCNJ12 |
IRK-2, IRK2, KCNJN1, Kir2.2, Kir2.2v, hIRK, hIRK1, hkir2.2x, kcnj12x |
-108.4kb | Anti-sense | Chr. 17 - 21376387 - 21419870 on NCBI Build GRCh38 |
Downstream | potassium voltage-gated channel subfamily J member 18 | KCNJ18 |
KIR2.6, TTPP2 |
162.4kb | Anti-sense | Chr. 17 - 21692523 - 21705700 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
