Assay Details
| Gene Symbol: PIGV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097053.1 | - | 1 | 948 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | AT-rich interaction domain 1A | ARID1A |
B120, BAF250, BAF250a, BM029, C1orf4, CSS2, ELD, MRD14, OSA1, P270, SMARCF1, hELD, hOSA1 |
-14.5kb | Sense | Chr. 1 - 26696031 - 26782110 on NCBI Build GRCh38 |
| Downstream | zinc finger DHHC-type containing 18 | ZDHHC18 |
DHHC-18, DHHC18 |
28.4kb | Sense | Chr. 1 - 26826710 - 26855720 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PIGV
Genomic Map