Assay Details
| Gene Symbol: DYNLRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097281.1 | - | 1 | 1650 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | itchy E3 ubiquitin protein ligase | ITCH |
ADMFD, AIF4, AIP4, NAPP1 |
-23.4kb | Sense | Chr. 20 - 34363235 - 34511773 on NCBI Build GRCh38 |
| Downstream | microtubule associated protein 1 light chain 3 alpha | MAP1LC3A |
ATG8E, LC3, LC3A, MAP1ALC3, MAP1BLC3 |
10.1kb | Sense | Chr. 20 - 34546823 - 34560345 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DYNLRB1
Genomic Map