Assay Details
| Gene Symbol: NAA35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011518903.2 | XP_011517205.1 | 20 | 5766 | 90 | |
| XM_011518904.2 | XP_011517206.1 | 17 | 5452 | 90 | ||
| XM_017015012.1 | XP_016870501.1 | 24 | 6022 | 90 | ||
| GenBank mRNA | AK097313.1 | - | 1 | 1748 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 27 pseudogene | LOC389765 | -181.0kb | Sense | Chr. 9 - 85806002 - 85842879 on NCBI Build GRCh38 | |
| Downstream | golgi membrane protein 1 | GOLM1 |
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3 |
368.0 | Anti-sense | Chr. 9 - 86026141 - 86100201 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NAA35
Genomic Map