Assay Details
| Gene Symbol: MKNK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038403.1 | 7 - 8 | 664 | 71 | ||
| GenBank mRNA | AK097731.1 | - | 7 - 8 | 664 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinocilin | KNCN |
Kino, L5 |
-16.5kb | Anti-sense | Chr. 1 - 46545638 - 46551657 on NCBI Build GRCh38 |
| Downstream | MOB kinase activator 3C | MOB3C |
MOB1E, MOBKL2C |
37.5kb | Anti-sense | Chr. 1 - 46607715 - 46616891 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000589.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20204553 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179054 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179033 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179024 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179046 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179026 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179048 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179058 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179035 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179061 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179047 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179040 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20204570 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179037 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179060 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179021 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179041 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20204573 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179043 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179031 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179044 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179028 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179022 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179056 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179057 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20204550 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179029 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179050 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20204535 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179045 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179034 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179030 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20204540 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20204571 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179023 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179042 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179032 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179049 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00179027 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 55897 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 571530 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 283701 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 239664 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 180462 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 133487 | AJ416091.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 133487 | BC085006.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 133487 | BC109254.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 133487 | BC110393.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 133487 | NM_133457.2 | refseq_mrna_human_NM |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00058679 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P35114382 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00058684 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35114382 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00058655 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00058679 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00058651 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P30152083 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00058676 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00058679 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P30152083 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P35114382 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01010047.hg.1 | TC01000589.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010047.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158466.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158466.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158466.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158466.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158466.hg.1 | TC0100008166.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | ENST00000602433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008166.hg.1 | NR_038403.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A000019 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A000019 | - | - |

MKNK1-AS1
Genomic Map