Assay Details
| Gene Symbol: XPOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017018748.1 | XP_016874237.1 | 25 | 4490 | 93 | |
| GenBank mRNA | AK098081.1 | - | 1 | 647 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein S11 pseudogene 6 | RPS11P6 | RPS11_2_1249 | -51.7kb | Sense | Chr. 12 - 64396010 - 64397000 on NCBI Build GRCh38 |
| Downstream | TANK binding kinase 1 | TBK1 |
FTDALS4, NAK, T2K |
937.0 | Sense | Chr. 12 - 64452060 - 64502119 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

XPOT
Genomic Map