Assay Details
| Gene Symbol: LOC105373434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC039108.1 | - | 1 | 755 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIR3681 host gene | MIR3681HG | -137.1kb | Sense | Chr. 2 - 12007116 - 12578348 on NCBI Build GRCh38 | |
| Downstream | tribbles pseudokinase 2 | TRIB2 |
C5FW, GS3955, TRB2 |
645.0 | Sense | Chr. 2 - 12716872 - 12742734 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105373434
Genomic Map