Assay Details
| Gene Symbol: GRIK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000830.4 | NP_000821.1 | 17 | 5737 | 103 | |
| GenBank mRNA | BC039447.1 | - | 1 | 350 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BACH1 intronic transcript 2 | BACH1-IT2 | -176.9kb | Anti-sense | Chr. 21 - 29372501 - 29373980 on NCBI Build GRCh38 | |
| Downstream | GRIK1 antisense RNA 2 | GRIK1-AS2 |
C21orf41, NCRNA00258 |
44.6kb | Anti-sense | Chr. 21 - 29596040 - 29630747 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GRIK1
Genomic Map