Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008529.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008529.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008529.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008529.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008529.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008529.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008529.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008529.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008529.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008529.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008529.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008529.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008529.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008529.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008529.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008529.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008529.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008529.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008529.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008529.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008529.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008529.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008529.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008529.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158451.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158451.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158451.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158451.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158451.hg.1 |
TC0500008529.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500158449.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158449.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158449.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158449.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158449.hg.1 |
TC0500008529.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500158450.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158450.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158450.hg.1 |
ENST00000564199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158450.hg.1 |
NR_109887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158450.hg.1 |
TC0500008529.hg.1 |
Clariom D TC |