Assay Details
Gene Symbol: LOC101928031 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC039486.1 | - | 1 - 2 | 55 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | dihydrofolate reductase pseudogene | LOC1720 | -665.0kb | Sense | Chr. 2 - 82856803 - 82857769 on NCBI Build GRCh38 | |
Downstream | FUN14 domain containing 2 pseudogene 2 | FUNDC2P2 | 767.2kb | Sense | Chr. 2 - 84290682 - 84292200 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
