Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011741.hg.1 |
ENST00000420350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011741.hg.1 |
ENST00000420350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011741.hg.1 |
ENST00000420350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011741.hg.1 |
NR_110668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011741.hg.1 |
NR_110668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011741.hg.1 |
NR_110668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100186496.hg.1 |
ENST00000420350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186496.hg.1 |
NR_110668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100186496.hg.1 |
ENST00000420350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186496.hg.1 |
NR_110668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100186496.hg.1 |
TC0100011741.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100186495.hg.1 |
ENST00000420350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186495.hg.1 |
NR_110668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100186495.hg.1 |
ENST00000420350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186495.hg.1 |
NR_110668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100186495.hg.1 |
TC0100011741.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011741.hg.1 |
ENST00000420350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011741.hg.1 |
ENST00000420350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011741.hg.1 |
ENST00000420350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011741.hg.1 |
NR_110668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011741.hg.1 |
NR_110668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011741.hg.1 |
NR_110668.1 |
Refseq |