Assay Details
| Gene Symbol: LINC01254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC040323.1 | - | 1 | 2853 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | VAMP associated protein A | VAPA |
VAP-33, VAP-A, VAP33, hVAP-33 |
-451.6kb | Anti-sense | Chr. 18 - 9913958 - 9960021 on NCBI Build GRCh38 |
| Downstream | APC down-regulated 1 | APCDD1 |
B7323, DRAPC1, FP7019, HHS, HTS, HYPT1 |
40.1kb | Anti-sense | Chr. 18 - 10454628 - 10488701 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LINC01254
Genomic Map