| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013727.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013727.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013727.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013727.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013727.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013727.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013727.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013727.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013727.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013727.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013727.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013727.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200200607.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200200607.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200200607.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200200607.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200200607.hg.1 |
TC0200013727.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200013727.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013727.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013727.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013727.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013727.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013727.hg.1 |
ENST00000430551.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013727.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013727.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013727.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013727.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013727.hg.1 |
NR_110113.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013727.hg.1 |
NR_110113.1 |
Refseq |