Assay Details
| Gene Symbol: LOC102724449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC041449.1 | - | 1 - 2 | 105 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC150051 | LOC150051 | -97.4kb | Anti-sense | Chr. 21 - 31559245 - 31560487 on NCBI Build GRCh38 | |
| Downstream |
superoxide dismutase 1, soluble |
SOD1 |
ALS, ALS1, HEL-S-44, IPOA, SOD, hSod1, homodimer |
122.0 | Anti-sense | Chr. 21 - 31659622 - 31668931 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC102724449
Genomic Map