Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008986.hg.1 |
ENST00000582251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008986.hg.1 |
ENST00000582251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008986.hg.1 |
ENST00000582251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008986.hg.1 |
NR_110764.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008986.hg.1 |
NR_110764.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008986.hg.1 |
NR_110764.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161672.hg.1 |
ENST00000582251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161672.hg.1 |
NR_110764.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161672.hg.1 |
TC1800008986.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800161672.hg.1 |
ENST00000582251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161672.hg.1 |
NR_110764.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161671.hg.1 |
ENST00000582251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161671.hg.1 |
NR_110764.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161671.hg.1 |
TC1800008986.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800161671.hg.1 |
ENST00000582251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161671.hg.1 |
NR_110764.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161674.hg.1 |
ENST00000582251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161674.hg.1 |
NR_110764.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161674.hg.1 |
TC1800008986.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800161674.hg.1 |
ENST00000582251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161674.hg.1 |
NR_110764.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008986.hg.1 |
ENST00000582251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008986.hg.1 |
ENST00000582251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008986.hg.1 |
ENST00000582251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008986.hg.1 |
NR_110764.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008986.hg.1 |
NR_110764.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008986.hg.1 |
NR_110764.1 |
Refseq |