Assay Details
| Gene Symbol: LOC101929555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110873.1 | 5 - 6 | 680 | 111 | ||
| GenBank mRNA | CR936713.1 | - | 5 - 6 | 680 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat and fibronectin type III domain containing 2 | LRFN2 |
FIGLER2, KIAA1246, SALM1 |
-291.4kb | Sense | Chr. 6 - 40391589 - 40587464 on NCBI Build GRCh38 |
| Downstream | unc-5 family C-terminal like | UNC5CL |
MUXA, ZUD |
143.8kb | Sense | Chr. 6 - 41026901 - 41039199 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101929555
Genomic Map