| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008769.hg.1 |
ENST00000438877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008769.hg.1 |
ENST00000438877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008769.hg.1 |
ENST00000438877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008769.hg.1 |
NR_111915.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008769.hg.1 |
NR_111915.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008769.hg.1 |
NR_111915.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008769.hg.1 |
NR_111915.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160912.hg.1 |
ENST00000438877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600160912.hg.1 |
NR_111915.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160912.hg.1 |
ENST00000438877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600160912.hg.1 |
NR_111915.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160912.hg.1 |
TC0600008769.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600008769.hg.1 |
ENST00000438877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008769.hg.1 |
ENST00000438877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008769.hg.1 |
ENST00000438877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008769.hg.1 |
NR_111915.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008769.hg.1 |
NR_111915.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008769.hg.1 |
NR_111915.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008769.hg.1 |
NR_111915.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160910.hg.1 |
NR_111915.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160910.hg.1 |
NR_111915.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160910.hg.1 |
TC0600008769.hg.1 |
Clariom D TC |