Assay Details
| Gene Symbol: MDM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130328.1 | - | 1 | 947 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta | PIK3C2B | C2-PI3K | -85.2kb | Anti-sense | Chr. 1 - 204422630 - 204494815 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105371692 | LOC105371692 | 12.7kb | Sense | Chr. 1 - 204559683 - 204628442 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MDM4
Genomic Map