Assay Details
| Gene Symbol: LOC101929454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109944.1 | 1 - 2 | 145 | 73 | ||
| GenBank mRNA | BC002741.2 | - | 1 - 2 | 145 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ANKH inorganic pyrophosphate transport regulator | ANKH |
ANK, CCAL2, CMDJ, CPPDD, HANK, MANK |
-371.9kb | Sense | Chr. 5 - 14704800 - 14871778 on NCBI Build GRCh38 |
| Downstream | F-box and leucine rich repeat protein 7 | FBXL7 |
FBL6, FBL7 |
233.6kb | Anti-sense | Chr. 5 - 15500196 - 15939796 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929454
Genomic Map