Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010182.hg.1 |
ENST00000511443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010182.hg.1 |
ENST00000511443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010182.hg.1 |
ENST00000511443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010182.hg.1 |
NR_109944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010182.hg.1 |
NR_109944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010182.hg.1 |
NR_109944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010182.hg.1 |
NR_109944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171272.hg.1 |
NR_109944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171272.hg.1 |
NR_109944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171272.hg.1 |
TC0500010182.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010182.hg.1 |
ENST00000511443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010182.hg.1 |
ENST00000511443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010182.hg.1 |
ENST00000511443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010182.hg.1 |
NR_109944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010182.hg.1 |
NR_109944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010182.hg.1 |
NR_109944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010182.hg.1 |
NR_109944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171273.hg.1 |
ENST00000511443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171273.hg.1 |
NR_109944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171273.hg.1 |
ENST00000511443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171273.hg.1 |
NR_109944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171273.hg.1 |
TC0500010182.hg.1 |
Clariom D TC |