Assay Details
Gene Symbol: LOC101929748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_136301.1 | 1 | 1731 | 86 | ||
GenBank mRNA | BC040975.2 | - | 1 | 1156 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | BICD cargo adaptor 2 | BICD2 |
SMALED2, bA526D8.1 |
-40.6kb | Sense | Chr. 9 - 92711363 - 92764841 on NCBI Build GRCh38 |
Downstream |
ankyrin repeat domain 19, pseudogene |
ANKRD19P |
ANKRD19, bA526D8.2 |
2.1kb | Anti-sense | Chr. 9 - 92809611 - 92838457 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
