Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008542.hg.1 |
ENST00000428285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008542.hg.1 |
ENST00000428285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008542.hg.1 |
ENST00000428285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008542.hg.1 |
ENST00000428285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008542.hg.1 |
NR_109955.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008542.hg.1 |
NR_109955.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008542.hg.1 |
NR_109955.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008542.hg.1 |
NR_109955.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159964.hg.1 |
ENST00000428285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159964.hg.1 |
NR_109955.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159964.hg.1 |
TC2000008542.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000159964.hg.1 |
ENST00000428285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159964.hg.1 |
NR_109955.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008542.hg.1 |
ENST00000428285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008542.hg.1 |
ENST00000428285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008542.hg.1 |
ENST00000428285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008542.hg.1 |
ENST00000428285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008542.hg.1 |
NR_109955.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008542.hg.1 |
NR_109955.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008542.hg.1 |
NR_109955.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008542.hg.1 |
NR_109955.1 |
Refseq |