Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009410.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009410.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009410.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009410.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009410.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009410.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009410.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009410.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009410.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009410.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009410.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009410.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164428.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164428.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164428.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164428.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164428.hg.1 |
TC0X00009410.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009410.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009410.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009410.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009410.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009410.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009410.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009410.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009410.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009410.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009410.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009410.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009410.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164429.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164429.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164429.hg.1 |
ENST00000431646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164429.hg.1 |
NR_110385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164429.hg.1 |
TC0X00009410.hg.1 |
Clariom D TC |