Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008579.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008579.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008579.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008579.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008579.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008579.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008579.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008579.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008579.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008579.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163054.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163054.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163054.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163054.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163054.hg.1 |
TC1200008579.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200008579.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008579.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008579.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008579.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008579.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008579.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008579.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008579.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008579.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008579.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163053.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163053.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163053.hg.1 |
ENST00000547633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163053.hg.1 |
NR_110095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163053.hg.1 |
TC1200008579.hg.1 |
Clariom D TC |