Assay Details
| Gene Symbol: LOC101927490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC043588.1 | - | 1 - 2 | 133 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105377622 | LOC105377622 | -9.6kb | Sense | Chr. 4 - 139600624 - 139612239 on NCBI Build GRCh38 | |
| Downstream | microsomal glutathione S-transferase 2 | MGST2 |
GST2, MGST-II |
42.5kb | Anti-sense | Chr. 4 - 139665725 - 139754546 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927490
Genomic Map