Assay Details
| Gene Symbol: LOC101928406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC044856.1 | - | 1 - 2 | 207 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dystrobrevin binding protein 1 | DTNBP1 |
BLOC1S8, DBND, HPS7, My031, SDY |
-331.9kb | Anti-sense | Chr. 6 - 15522801 - 15663058 on NCBI Build GRCh38 |
| Downstream | myosin regulatory light chain interacting protein | MYLIP |
IDOL, MIR |
128.6kb | Sense | Chr. 6 - 16129086 - 16151015 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101928406
Genomic Map