Assay Details
| Gene Symbol: LOC105378910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC044921.2 | - | 1 | 1756 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein phosphatase, Mg2+/Mn2+ dependent 1J |
PPM1J |
PP2CZ, PP2Czeta, PPP2CZ |
-344.0 | Anti-sense | Chr. 1 - 112709994 - 112715328 on NCBI Build GRCh38 |
| Downstream |
family with sequence similarity 19 member A3, C-C motif chemokine like |
FAM19A3 |
TAFA-3, TAFA3 |
1.2kb | Sense | Chr. 1 - 112719003 - 112727235 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105378910
Genomic Map