Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX537630.1 | - | 1 | 1853 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 4677 | MIR4677 | mir-4677 | -150.1kb | Sense | Chr. 1 - 243346176 - 243346255 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC339529 | LOC339529 | 417.3kb | Sense | Chr. 1 - 243917402 - 244047317 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
