Assay Details
| Gene Symbol: CD34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX640941.1 | - | 1 | 3849 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 1 open reading frame 132 | C1orf132 | -11.7kb | Sense | Chr. 1 - 207801518 - 207822703 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105372883 | LOC105372883 | 77.0kb | Anti-sense | Chr. 1 - 207962873 - 207968142 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |

CD34
Genomic Map