Assay Details
Gene Symbol: TUSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017013861.1 | XP_016869350.1 | 10 | 3652 | 86 | |
GenBank mRNA | BX641112.1 | - | 1 | 1852 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | sarcoglycan zeta | SGCZ | ZSG1 | -522.7kb | Anti-sense | Chr. 8 - 14084835 - 15238283 on NCBI Build GRCh38 |
Downstream | macrophage scavenger receptor 1 | MSR1 |
CD204, SCARA1, SR-A, SRA, phSR1, phSR2 |
343.2kb | Anti-sense | Chr. 8 - 16107878 - 16192791 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
