| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000436187.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000436187.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000436187.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000436187.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000436187.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000436187.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011660.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200184667.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184667.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200184667.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184667.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200184667.hg.1 |
TC0200011660.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200184669.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184669.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200184669.hg.1 |
ENST00000454224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200184669.hg.1 |
NR_110257.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200184669.hg.1 |
TC0200011660.hg.1 |
Clariom D TC |