Assay Details
| Gene Symbol: GLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX641148.1 | - | 1 | 254 | 131 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NGFI-A binding protein 1 | NAB1 | -239.7kb | Sense | Chr. 2 - 190648860 - 190692766 on NCBI Build GRCh38 | |
| Downstream | signal transducer and activator of transcription 1 | STAT1 |
CANDF7, IMD31A, IMD31B, IMD31C, ISGF-3, STAT91 |
33.7kb | Anti-sense | Chr. 2 - 190969036 - 191014250 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000412247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000412247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000412247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000412247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000412247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000412247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000468352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000468352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000468352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000468352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000468352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000468352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000469774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000469774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000469774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000469774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000469774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000469774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000471443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000471443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000471443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000471443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000471443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000495444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000495444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000495444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000495444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000495444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000496170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000496170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000496170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000496170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000496170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000629173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |

GLS
Genomic Map