Assay Details
| Gene Symbol: LOC101928377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX647685.1 | - | 1 | 3753 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1220 | LINC01220 | -127.3kb | Anti-sense | Chr. 14 - 75294404 - 75296410 on NCBI Build GRCh38 | |
| Downstream | Jun dimerization protein 2 | JDP2 | JUNDM2 | 65.0 | Anti-sense | Chr. 14 - 75426685 - 75472701 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101928377
Genomic Map