Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163435.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163435.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163435.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163435.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163435.hg.1 |
TC0400009233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400163433.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163433.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163433.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163433.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163433.hg.1 |
TC0400009233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400163436.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163436.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163436.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163436.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163436.hg.1 |
TC0400009233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009233.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163434.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163434.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163434.hg.1 |
ENST00000507838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163434.hg.1 |
NR_121676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163434.hg.1 |
TC0400009233.hg.1 |
Clariom D TC |