Assay Details
| Gene Symbol: IAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123278.1 | - | 1 | 849 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cleavage and polyadenylation specific factor 3 | CPSF3 |
CPSF-73, CPSF73 |
-24.6kb | Sense | Chr. 2 - 9423537 - 9473110 on NCBI Build GRCh38 |
| Downstream | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta | YWHAQ |
14-3-3, 1C5, HS1 |
83.4kb | Anti-sense | Chr. 2 - 9583967 - 9631055 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

IAH1
Genomic Map