Assay Details
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005252320.3 | XP_005252377.1 | 7 | 2592 | 95 | |
GenBank mRNA | AK123739.1 | - | 1 | 1353 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | N-acetylneuraminate synthase | NANS |
HEL-S-100, SAS, SEMDG |
Anti-sense | Chr. 9 - 98056677 - 98083083 on NCBI Build GRCh38 | |
Downstream | coronin 2A | CORO2A |
CLIPINB, IR10, WDR2 |
37.1kb | Sense | Chr. 9 - 98120975 - 98192674 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
