Assay Details
Gene Symbol: SERTAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK124032.1 | - | 1 | 1248 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC339807 | LOC339807 | -132.0kb | Anti-sense | Chr. 2 - 64607312 - 64616482 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101927438 | LOC101927438 | 95.0kb | Sense | Chr. 2 - 64846130 - 64863631 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
