Assay Details
Gene Symbol: LOC100288860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK124080.1 | - | 1 | 952 | 85 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | T62183.1 | - | 54 | Chr. - - [-] on Build | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | potassium channel tetramerization domain containing 15 | KCTD15 | -90.6kb | Sense | Chr. 19 - 33795540 - 33815761 on NCBI Build GRCh38 | |
Downstream |
LSM14A, mRNA processing body assembly factor |
LSM14A |
C19orf13, FAM61A, RAP55, RAP55A |
264.1kb | Sense | Chr. 19 - 34172428 - 34229515 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569457 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569471 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942965 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569462 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569498 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17132102 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569469 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942943 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942953 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569504 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569458 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942963 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569474 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942950 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569490 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942941 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17132095 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942957 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942973 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942931 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569453 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942947 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569500 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942948 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942971 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942964 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569491 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942975 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942935 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569494 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942955 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17132084 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569464 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569454 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942954 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569449 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942974 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942959 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569493 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942976 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569480 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942951 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942934 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942972 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31569489 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10942969 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 201957 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P80610 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 31323 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132095 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942961 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942954 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942970 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942973 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942975 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569449 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569507 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942972 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569454 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942957 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942956 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942942 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942958 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569503 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942969 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942938 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942964 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942967 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569496 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569491 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942937 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942962 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569500 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569453 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569470 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569465 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569460 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942974 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942935 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569464 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569492 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569456 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942941 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942934 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942948 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942936 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942959 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569474 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569458 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942949 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569490 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942952 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942960 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942945 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942951 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942933 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569463 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942976 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569461 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132086 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942955 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569493 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942966 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569471 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569498 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569504 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942939 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132083 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569472 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569448 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942963 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569469 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569451 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569480 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942944 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942947 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942968 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569466 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942940 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942971 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942946 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942932 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942950 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569462 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569506 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132102 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569489 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132088 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569494 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942965 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132085 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569467 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942931 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942953 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569457 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132084 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942943 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942969 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942954 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942945 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942948 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569472 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942972 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942940 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942933 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942946 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569448 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569463 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132088 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942935 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942964 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942953 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942936 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569456 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942955 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569465 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569461 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569454 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569466 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569504 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569507 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569480 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132085 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569490 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569467 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942942 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569453 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942931 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942961 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569457 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942941 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569494 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942949 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569498 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569491 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942957 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942960 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569449 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569451 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569458 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132102 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132083 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569460 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569469 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942962 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569503 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942976 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569471 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942959 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569493 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132095 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942963 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942938 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569506 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942956 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942932 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942937 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569462 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942939 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942958 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569496 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132086 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942965 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942975 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942971 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569492 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942944 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942973 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942966 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942950 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569489 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942952 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942951 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942934 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569474 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569464 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942970 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942947 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942943 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942968 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569470 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942967 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10942974 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31569500 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17132084 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P80610 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P80610 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P80602 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 239127_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1900007789.hg.1 | ENST00000610908.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007789.hg.1 | ENST00000610908.1 | Ensembl |
Human Genome U95 Set | 85544_at | - | - |
Human Genome U95 Set | 85544_PM_at | - | - |
Human Genome U133 Set | 239127_PM_at | - | - |
Human Genome U133 Set | 239127_at | - | - |
