Assay Details
| Gene Symbol: USB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204911.1 | NP_001191840.1 | 4 | 1457 | 72 | |
| GenBank mRNA | AK124443.1 | - | 1 | 1251 | 72 | |
| BF733890.1 | - | 1 | 72 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 319 | ZNF319 | ZFP319 | -10.0kb | Anti-sense | Chr. 16 - 57994668 - 58000691 on NCBI Build GRCh38 |
| Downstream | matrix metallopeptidase 15 | MMP15 |
MMP-15, MT2-MMP, MT2MMP, MTMMP2, SMCP-2 |
11.8kb | Sense | Chr. 16 - 58025566 - 58046901 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000499.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000564387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000564387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000564387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000564387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000564387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000564387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |

USB1
Genomic Map