Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011191.hg.1 |
ENST00000624850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008218.hg.1 |
ENST00000608229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012623.hg.1 |
ENST00000527533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012623.hg.1 |
ENST00000527533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012623.hg.1 |
ENST00000527774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012623.hg.1 |
ENST00000527774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012623.hg.1 |
ENST00000527774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012623.hg.1 |
ENST00000527533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012623.hg.1 |
ENST00000527533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012623.hg.1 |
ENST00000527774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012623.hg.1 |
ENST00000527774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012623.hg.1 |
ENST00000527774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011191.hg.1 |
ENST00000624850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008218.hg.1 |
ENST00000608229.1 |
Ensembl |