Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chordin like 1 | CHRDL1 |
CHL, MGC1, MGCN, NRLN1, VOPT, dA141H5.1 |
-426.6kb | Anti-sense | Chr. X - 110673856 - 110796058 on NCBI Build GRCh38 |
| Downstream | calpain 6 | CAPN6 |
CANPX, CAPNX, CalpM, DJ914P14.1 |
17.7kb | Anti-sense | Chr. X - 111245099 - 111270546 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PAK3
Genomic Map