Assay Details
Gene Symbol: CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK125081.1 | - | 1 | 1752 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | LY75-CD302 readthrough | LY75-CD302 | Sense | Chr. 2 - 159768628 - 159904756 on NCBI Build GRCh38 | ||
Downstream | lymphocyte antigen 75 | LY75 |
CD205, CLEC13B, DEC-205, GP200-MR6, LY-75 |
5.1kb | Sense | Chr. 2 - 159803355 - 159904756 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
