| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000448324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000448324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000448726.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000448726.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000448726.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370353.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000370354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000448324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000448324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000448726.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000448726.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000448726.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
ENST00000538575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NM_024082.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008735.hg.1 |
NR_033262.1 |
Refseq |