| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
NR_111905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
NR_111905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
NR_111905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
NR_111905.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012378.hg.1 |
NR_111905.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000366099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
ENST00000444433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
NR_111905.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
NR_111905.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
NR_111905.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
NR_111905.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012378.hg.1 |
NR_111905.1 |
Refseq |