Assay Details
| Gene Symbol: CNTN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125798.1 | - | 1 - 2 | 126 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1266 | LINC01266 | -162.1kb | Sense | Chr. 3 - 592105 - 846015 on NCBI Build GRCh38 | |
| Downstream | contactin 4 | CNTN4 |
AXCAM, BIG-2 |
1.1mb | Sense | Chr. 3 - 2098771 - 3059080 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

CNTN6
Genomic Map