Assay Details
| Gene Symbol: LOC100505635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125892.1 | - | 2 - 3 | 3261 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 6 | KIF6 |
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1 |
-156.3kb | Sense | Chr. 6 - 39335100 - 39725405 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC100505635
Genomic Map